Assay Details
| Gene Symbol: Abcg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104382.1 | 1 - 2 | 177 | 67 | ||
| GenBank mRNA | AY196215.1 | - | 1 - 2 | 167 | 67 | |
| AY196216.1 | - | 1 - 2 | 167 | 67 | ||
| BC138484.1 | - | 1 - 2 | 144 | 8861504 | 67 | |
| BC145293.1 | - | 1 - 2 | 144 | 9053847 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcg8
Genomic Map