Assay Details
| Gene Symbol: Cox7a2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK169619.1 | - | 2 - 3 | 270 | 67 | |
| AK216072.1 | - | 1 - 2 | 133 | 67 | ||
| BC004715.1 | - | 2 - 3 | 222 | 3582142 | 67 | |
| BC085296.1 | - | 2 - 3 | 227 | 5337317 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cox7a2l
Genomic Map