Assay Details
| Gene Symbol: Mta3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171052.1 | NP_001164523.1 | 10 - 11 | 1062 | 108 | |
| NM_001171053.1 | NP_001164524.1 | 10 - 11 | 1062 | 108 | ||
| NM_001171054.1 | NP_001164525.1 | 10 - 11 | 1062 | 108 | ||
| NM_054082.2 | NP_473423.1 | 10 - 11 | 1059 | 108 | ||
| GenBank mRNA | AK129318.1 | - | 10 - 11 | 991 | 108 | |
| AK145942.1 | - | 10 - 11 | 1007 | 108 | ||
| AK146721.1 | - | 10 - 11 | 1015 | 108 | ||
| AK146905.1 | - | 10 - 11 | 1015 | 108 | ||
| AK147197.1 | - | 10 - 11 | 1005 | 108 | ||
| AK154856.1 | - | 10 - 11 | 1002 | 108 | ||
| AK158866.1 | - | 9 - 10 | 925 | 108 | ||
| BC022124.1 | - | 10 - 11 | 1026 | 5097283 | 108 | |
| CA751287.1 | - | 4 - 5 | 402 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mta3
Genomic Map