Assay Details
| Gene Symbol: Strn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011500.2 | NP_035630.2 | 8 - 9 | 1118 | 126 | |
| GenBank mRNA | AJ223777.1 | - | 8 - 9 | 1045 | 126 | |
| AK135684.1 | - | 3 - 4 | 310 | 126 | ||
| AK136032.1 | - | 6 - 7 | 767 | 126 | ||
| BC090968.1 | - | 8 - 9 | 1118 | 30646517 | 126 | |
| BC150727.1 | - | 8 - 9 | 1086 | 9087638 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Strn
Genomic Map