Assay Details
| Gene Symbol: Fer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037997.3 | NP_001033086.2 | 7 - 8 | 1050 | 81 | |
| NM_001286415.1 | NP_001273344.1 | 6 - 7 | 878 | 81 | ||
| GenBank mRNA | AF286537.1 | - | 6 - 7 | 916 | 81 | |
| AK082799.1 | - | 4 - 5 | 546 | 81 | ||
| AK157828.1 | - | 7 - 8 | 1017 | 81 | ||
| AK165973.1 | - | 7 - 8 | 1051 | 81 | ||
| BC058100.1 | - | 6 - 7 | 878 | 6813507 | 81 | |
| U76762.1 | - | 6 - 7 | 983 | 81 | ||
More Information
Set Membership: |
|

Fer
Genomic Map