Assay Details
| Gene Symbol: Fer | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037997.3 | NP_001033086.2 | 9 - 10 | 1373 | 77 | |
| NM_001286415.1 | NP_001273344.1 | 8 - 9 | 1201 | 77 | ||
| GenBank mRNA | AF286537.1 | - | 8 - 9 | 1239 | 77 | |
| AK082799.1 | - | 6 - 7 | 869 | 77 | ||
| AK157828.1 | - | 9 - 10 | 1340 | 77 | ||
| BC051249.1 | - | 1 - 2 | 117 | 77 | ||
| BC058100.1 | - | 8 - 9 | 1201 | 6813507 | 77 | |
| U76762.1 | - | 8 - 9 | 1306 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fer
Genomic Map