Assay Details
| Gene Symbol: Ephb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168296.1 | NP_001161768.1 | 3 - 4 | 1158 | 80 | |
| NM_173447.3 | NP_775623.3 | 3 - 4 | 1158 | 80 | ||
| GenBank mRNA | AK033966.1 | - | 3 - 4 | 1158 | 80 | |
| AK036148.1 | - | 3 - 4 | 846 | 80 | ||
| AK036211.1 | - | 3 - 4 | 1157 | 80 | ||
| AK039530.1 | - | 3 - 4 | 1159 | 80 | ||
| AK135018.1 | - | 3 - 4 | 1124 | 80 | ||
| BC057301.1 | - | 3 - 4 | 1098 | 6821724 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ephb1
Genomic Map