Assay Details
| Gene Symbol: Manf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029103.3 | NP_083379.2 | 1 - 2 | 118 | 90 | |
| GenBank mRNA | AB009291.1 | - | 1 - 2 | 92 | 90 | |
| AK014338.1 | - | 1 - 2 | 96 | 90 | ||
| AK131997.1 | - | 1 - 2 | 119 | 90 | ||
| AK165646.1 | - | 1 - 2 | 94 | 90 | ||
| BC022761.1 | - | 1 - 2 | 126 | 90 | ||
| BC038901.1 | - | 1 - 2 | 106 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Manf
Genomic Map