Assay Details
| Gene Symbol: Ackr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145700.2 | NP_663746.2 | 2 - 3 | 493 | 158 | |
| GenBank mRNA | AK042430.1 | - | 1 - 2 | 164 | 158 | |
| AY072796.1 | - | 1 - 2 | 744 | 158 | ||
| AY072938.1 | - | 2 - 3 | 785 | 158 | ||
| BC140999.1 | - | 2 - 3 | 493 | 9055611 | 158 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK042430.1, AY072796.1, AY072938.1, BC140999.1, NM_145700.2 transcripts.
Set Membership: |
|

Ackr4
Genomic Map