Assay Details
| Gene Symbol: Kdm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172132.2 | NP_742144.1 | 10 - 11 | 1400 | 117 | |
| GenBank mRNA | AK078074.1 | - | 10 - 11 | 1393 | 117 | |
| AK129233.1 | - | 10 - 11 | 1359 | 117 | ||
| AK141879.1 | - | 10 - 11 | 1377 | 117 | ||
| BC007145.1 | - | 10 - 11 | 1370 | 3490671 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdm4b
Genomic Map