Assay Details
| Gene Symbol: Apool | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012398.1 | - | 1 - 2 | 73 | 63 | |
| AK012804.1 | - | 1 - 2 | 65 | 63 | ||
| AK028953.1 | - | 1 - 2 | 88 | 63 | ||
| AK079141.1 | - | 1 - 2 | 78 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK012804.1 transcripts.
Set Membership: |
|

Apool
Genomic Map