Assay Details
| Gene Symbol: Apool | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026565.2 | NP_080841.1 | 2 - 3 | 136 | 116 | |
| GenBank mRNA | BC024334.1 | - | 2 - 3 | 136 | 5036743 | 116 |
More Information
Updated Mapping Information
Set Membership: |
|

Apool
Genomic Map