Assay Details
| Gene Symbol: Klhdc8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030075.2 | NP_084351.1 | 4 - 5 | 558 | 69 | |
| GenBank mRNA | AK019849.1 | - | 4 - 5 | 558 | 69 | |
| AK048891.1 | - | 1 - 2 | 1560 | 69 | ||
| AK087143.1 | - | 4 - 5 | 544 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK019849.1, NM_030075.2 transcripts.
Set Membership: |
|

Klhdc8b
Genomic Map