Assay Details
| Gene Symbol: Klhdc8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030075.2 | NP_084351.1 | 8 - 9 | 1786 | 85 | |
| GenBank mRNA | AK019849.1 | - | 8 - 9 | 1784 | 85 | |
| AK154428.1 | - | 8 - 9 | 1887 | 85 | ||
| BC024793.1 | - | 1 - 2 | 1191 | 85 | ||
| BC048918.1 | - | 3 - 4 | 749 | 6309434 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klhdc8b
Genomic Map