Assay Details
| Gene Symbol: Klhdc8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030075.2 | NP_084351.1 | 2 - 3 | 213 | 66 | |
| GenBank mRNA | AK019849.1 | - | 2 - 3 | 213 | 66 | |
| AK087143.1 | - | 2 - 3 | 199 | 66 | ||
| AK154428.1 | - | 2 - 3 | 351 | 66 | ||
| AK161517.1 | - | 2 - 3 | 661 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK019849.1, AK154428.1, NM_030075.2 transcripts.
Set Membership: |
|

Klhdc8b
Genomic Map