Assay Details
| Gene Symbol: Abhd14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB073619.1 | - | 3 - 4 | 278 | 60 | |
| BC006879.1 | - | 3 - 4 | 287 | 2647263 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB073619.1, BC006879.1 transcripts.
Set Membership: |
|

Abhd14a
Genomic Map