Assay Details
| Gene Symbol: Taf9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001176.2 | NP_001001176.2 | 3 - 4 | 481 | 73 | |
| NM_001167988.1 | NP_001161460.1 | 4 - 5 | 419 | 73 | ||
| GenBank mRNA | AK131710.1 | - | 4 - 5 | 420 | 73 | |
| AK135097.1 | - | 3 - 4 | 478 | 73 | ||
| BC051635.1 | - | 3 - 4 | 279 | 73 | ||
| BC066223.1 | - | 3 - 4 | 272 | 30468965 | 73 | |
| CA559941.1 | - | 3 - 4 | 481 | 73 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Taf9b
Genomic Map