Assay Details
| Gene Symbol: Gpr108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF376726.1 | - | 12 - 13 | 1314 | 66 | |
| AK019012.1 | - | 3 - 4 | 288 | 66 | ||
| AK047364.1 | - | 12 - 13 | 1290 | 66 | ||
| AK082797.1 | - | 12 - 13 | 1339 | 66 | ||
| AK159313.1 | - | 12 - 13 | 1359 | 66 | ||
| AK170948.1 | - | 11 - 12 | 1442 | 66 | ||
| AK171030.1 | - | 11 - 12 | 1442 | 66 | ||
| AX775895.1 | - | 12 - 13 | 1371 | 66 | ||
| AX775897.1 | - | 12 - 13 | 1314 | 66 | ||
| BC016104.1 | - | 12 - 13 | 1340 | 4920840 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK019012.1 transcripts.
Set Membership: |
|

Gpr108
Genomic Map