Assay Details
| Gene Symbol: Gpr108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF376726.1 | - | 14 - 15 | 1408 | 72 | |
| AK019012.1 | - | 5 - 6 | 382 | 72 | ||
| AK047364.1 | - | 14 - 15 | 1384 | 72 | ||
| AK082797.1 | - | 14 - 15 | 1433 | 72 | ||
| AK159313.1 | - | 14 - 15 | 1453 | 72 | ||
| AK170948.1 | - | 13 - 14 | 1536 | 72 | ||
| AK171030.1 | - | 13 - 14 | 1536 | 72 | ||
| AX775895.1 | - | 14 - 15 | 1465 | 72 | ||
| AX775897.1 | - | 14 - 15 | 1408 | 72 | ||
| BC016104.1 | - | 14 - 15 | 1434 | 4920840 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK019012.1 transcripts.
Set Membership: |
|

Gpr108
Genomic Map