Assay Details
| Gene Symbol: Gpr108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF376726.1 | - | 9 - 10 | 993 | 63 | |
| AK047364.1 | - | 9 - 10 | 969 | 63 | ||
| AK082797.1 | - | 9 - 10 | 1019 | 63 | ||
| AK159313.1 | - | 9 - 10 | 1038 | 63 | ||
| AK170948.1 | - | 9 - 10 | 1021 | 63 | ||
| AK171030.1 | - | 9 - 10 | 1021 | 63 | ||
| AK215617.1 | - | 1 - 2 | 81 | 63 | ||
| AX775895.1 | - | 9 - 10 | 1050 | 63 | ||
| AX775897.1 | - | 9 - 10 | 993 | 63 | ||
| BC016104.1 | - | 9 - 10 | 1019 | 4920840 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr108
Genomic Map