Assay Details
| Gene Symbol: Khsrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010613.3 | NP_034743.3 | 14 - 15 | 1708 | 82 | |
| GenBank mRNA | AK018152.1 | - | 2 - 3 | 167 | 82 | |
| AK052249.1 | - | 10 - 11 | 1160 | 82 | ||
| AK077415.1 | - | 14 - 15 | 1678 | 82 | ||
| AK156541.1 | - | 14 - 15 | 1686 | 82 | ||
| BC064454.1 | - | 3 - 4 | 289 | 82 | ||
| BC108414.1 | - | 8 - 9 | 1009 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK018152.1 transcripts.
Set Membership: |
|

Khsrp
Genomic Map