Assay Details
| Gene Symbol: Tm9sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080556.3 | NP_542123.3 | 10 - 11 | 1335 | 109 | |
| GenBank mRNA | AK010325.1 | - | 10 - 11 | 1292 | 109 | |
| AK030340.1 | - | 10 - 11 | 1266 | 109 | ||
| AK048012.1 | - | 10 - 11 | 1254 | 109 | ||
| AK050338.1 | - | 10 - 11 | 1282 | 109 | ||
| AK075720.1 | - | 10 - 11 | 1335 | 109 | ||
| AK088905.1 | - | 10 - 11 | 1307 | 109 | ||
| AK159757.1 | - | 10 - 11 | 1280 | 109 | ||
| AK179255.1 | - | 1 - 2 | 137 | 109 | ||
| BC003862.1 | - | 10 - 11 | 1274 | 3488326 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tm9sf2
Genomic Map