Assay Details
| Gene Symbol: Vps51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081041.1 | NP_001074510.1 | 1 - 2 | 237 | 82 | |
| GenBank mRNA | AK014229.1 | - | 1 - 2 | 251 | 82 | |
| AK137155.1 | - | 1 - 2 | 247 | 82 | ||
| AK146211.1 | - | 1 - 2 | 246 | 82 | ||
| BC034452.1 | - | 1 - 2 | 253 | 82 | ||
| BC051479.1 | - | 1 - 2 | 243 | 82 | ||
| BC118930.2 | - | 1 - 2 | 252 | 40090408 | 82 | |
| BC118931.1 | - | 1 - 2 | 252 | 40090409 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vps51
Genomic Map