Assay Details
| Gene Symbol: Crim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015800.3 | NP_056615.1 | 4 - 5 | 1386 | 62 | |
| GenBank mRNA | AF168680.1 | - | 4 - 5 | 838 | 62 | |
| AK053197.1 | - | 2 - 3 | 329 | 62 | ||
| BC100348.1 | - | 4 - 5 | 1171 | 30521148 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crim1
Genomic Map