Assay Details
| Gene Symbol: Camkmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028576.2 | NP_082852.1 | 3 - 4 | 400 | 82 | |
| GenBank mRNA | AK155246.1 | - | 3 - 4 | 401 | 82 | |
| BC138272.1 | - | 3 - 4 | 548 | 8861291 | 82 | |
| BC138273.1 | - | 3 - 4 | 548 | 8861292 | 82 | |
| Gene Symbol: Gnao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK147783.1 | - | 437 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Camkmt
Genomic Map