Assay Details
| Gene Symbol: Camkmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028576.2 | NP_082852.1 | 7 - 8 | 649 | 79 | |
| GenBank mRNA | AK155246.1 | - | 7 - 8 | 650 | 79 | |
| BC138272.1 | - | 7 - 8 | 797 | 8861291 | 79 | |
| BC138273.1 | - | 7 - 8 | 797 | 8861292 | 79 | |
| Gene Symbol: Gnao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK147783.1 | - | 687 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Camkmt
Genomic Map