Assay Details
| Gene Symbol: Nudt12os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015703.1 | - | 1 - 2 | 263 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015703.1 transcripts.
Set Membership: |
|

Nudt12os
Genomic Map