Assay Details
| Gene Symbol: Nudt12os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015703.1 | - | 3 - 4 | 826 | 67 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003601 | AK015703.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 400546 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890736 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939952 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 796549 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 328447 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 910494 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367414 | AF163846.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367414 | AF208031.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367414 | AK033512.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367414 | AK085602.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367414 | BC003845.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367414 | NM_019810.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174043 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 586227 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278445 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 152731 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154868 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003601 | AK015703.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08816856 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08816843 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03343272 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154868 | - | - |
| Whole Mouse Genome G4122A | A_51_P154818 | AK015703.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154818 | BC057657.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154818 | AK083846.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154818 | AK147867.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P154818 | NM_026497.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100171 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154868 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08816856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267860 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267829 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267833 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13338467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267827 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267862 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267857 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13400039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267812 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267809 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13484679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13469973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267828 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267820 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267824 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13400037 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31267840 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10446343 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446340 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446342 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10446337 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432424_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10446341 | - | - |
| Mouse Genome 430 2.0 Array | 1432424_at | - | - |
| Mouse Genome 430 2.0 Array | 1432424_PM_at | - | - |
| Mouse Expression Set 430 | 1432424_at | - | - |
| Mouse Expression Set 430 | 1432424_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432424_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432424_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104150133 | - | - |

Nudt12os
Genomic Map