Assay Details
| Gene Symbol: Clip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271483.1 | NP_001258412.1 | 12 - 13 | 1677 | 78 | |
| NM_001271484.1 | NP_001258413.1 | 12 - 13 | 1762 | 78 | ||
| NM_030179.3 | NP_084455.2 | 12 - 13 | 1665 | 78 | ||
| NM_175378.1 | NP_780587.1 | 11 - 12 | 1621 | 78 | ||
| NR_073185.1 | 12 - 13 | 1817 | 78 | |||
| GenBank mRNA | AK017914.1 | - | 9 - 10 | 1253 | 78 | |
| AK019516.1 | - | 12 - 13 | 1678 | 78 | ||
| AK029962.1 | - | 12 - 13 | 1818 | 78 | ||
| AK075758.1 | - | 11 - 12 | 1612 | 78 | ||
| AK161301.1 | - | 12 - 13 | 1752 | 78 | ||
| AK163644.1 | - | 5 - 6 | 605 | 78 | ||
| BC011280.1 | - | 5 - 6 | 529 | 78 | ||
| BC035226.1 | - | 12 - 13 | 1665 | 4920788 | 78 | |
More Information
Set Membership: |
|

Clip4
Genomic Map