Assay Details
| Gene Symbol: Clip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271483.1 | NP_001258412.1 | 5 - 6 | 678 | 92 | |
| NM_001271484.1 | NP_001258413.1 | 5 - 6 | 763 | 92 | ||
| NM_030179.3 | NP_084455.2 | 5 - 6 | 666 | 92 | ||
| NM_175378.1 | NP_780587.1 | 5 - 6 | 763 | 92 | ||
| NR_073185.1 | 5 - 6 | 763 | 92 | |||
| GenBank mRNA | AK017914.1 | - | 2 - 3 | 253 | 92 | |
| AK018895.1 | - | 5 - 6 | 756 | 92 | ||
| AK019516.1 | - | 5 - 6 | 679 | 92 | ||
| AK029962.1 | - | 5 - 6 | 764 | 92 | ||
| AK075758.1 | - | 5 - 6 | 754 | 92 | ||
| AK161301.1 | - | 5 - 6 | 753 | 92 | ||
| BC035226.1 | - | 5 - 6 | 666 | 4920788 | 92 | |
More Information
Set Membership: |
|

Clip4
Genomic Map