Assay Details
| Gene Symbol: Galnt14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027864.2 | NP_082140.2 | 12 - 13 | 1623 | 73 | |
| GenBank mRNA | AB175681.1 | - | 12 - 13 | 1231 | 73 | |
| AK049598.1 | - | 12 - 13 | 1569 | 73 | ||
| BC037716.1 | - | 6 - 7 | 602 | 73 | ||
| BC117800.1 | - | 12 - 13 | 1407 | 40092267 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt14
Genomic Map