Assay Details
| Gene Symbol: Galnt14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027864.2 | NP_082140.2 | 7 - 8 | 1131 | 109 | |
| GenBank mRNA | AB175681.1 | - | 7 - 8 | 739 | 109 | |
| AK049598.1 | - | 7 - 8 | 1077 | 109 | ||
| AK078292.1 | - | 7 - 8 | 744 | 109 | ||
| BC037716.1 | - | 1 - 2 | 110 | 109 | ||
| BC117800.1 | - | 7 - 8 | 915 | 40092267 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galnt14
Genomic Map