Assay Details
| Gene Symbol: Txnrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042513.1 | NP_001035978.1 | 12 - 13 | 1438 | 159 | |
| NM_001042514.1 | NP_001035979.1 | 11 - 12 | 1526 | 159 | ||
| NM_001042523.1 | NP_001035988.1 | 12 - 13 | 1655 | 159 | ||
| NM_015762.2 | NP_056577.2 | 11 - 12 | 1331 | 159 | ||
| GenBank mRNA | AB027565.1 | - | 11 - 12 | 1358 | 159 | |
| AF333036.1 | - | 12 - 13 | 1655 | 159 | ||
| AK146125.1 | - | 11 - 12 | 1332 | 159 | ||
| AK151927.1 | - | 12 - 13 | 1438 | 159 | ||
| AK168356.1 | - | 12 - 13 | 1439 | 159 | ||
| BC020274.1 | - | 11 - 12 | 1293 | 159 | ||
| BC037643.1 | - | 11 - 12 | 1611 | 5059831 | 159 | |
More Information
Updated Mapping Information
Set Membership: |
|

Txnrd1
Genomic Map