Assay Details
| Gene Symbol: Txnrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042513.1 | NP_001035978.1 | 6 - 7 | 664 | 85 | |
| NM_001042514.1 | NP_001035979.1 | 5 - 6 | 752 | 85 | ||
| NM_001042523.1 | NP_001035988.1 | 6 - 7 | 881 | 85 | ||
| NM_015762.2 | NP_056577.2 | 5 - 6 | 557 | 85 | ||
| GenBank mRNA | AB027565.1 | - | 5 - 6 | 584 | 85 | |
| AF333036.1 | - | 6 - 7 | 881 | 85 | ||
| AK011902.1 | - | 6 - 7 | 665 | 85 | ||
| AK146125.1 | - | 5 - 6 | 558 | 85 | ||
| AK149625.1 | - | 6 - 7 | 665 | 85 | ||
| AK151927.1 | - | 6 - 7 | 665 | 85 | ||
| AK168356.1 | - | 6 - 7 | 665 | 85 | ||
| AK212800.1 | - | 2 - 3 | 255 | 85 | ||
| BC037643.1 | - | 5 - 6 | 837 | 5059831 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Txnrd1
Genomic Map