Assay Details
| Gene Symbol: Aff3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290814.1 | NP_001277743.1 | 7 - 8 | 1143 | 95 | |
| GenBank mRNA | AK014322.1 | - | 4 - 5 | 451 | 95 | |
| BC052061.1 | - | 8 - 9 | 1554 | 95 | ||
More Information
Set Membership: |
|

Aff3
Genomic Map