Assay Details
| Gene Symbol: Aff3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290814.1 | NP_001277743.1 | 8 - 9 | 1192 | 78 | |
| GenBank mRNA | AK014322.1 | - | 5 - 6 | 500 | 78 | |
| BC052061.1 | - | 9 - 10 | 1603 | 78 | ||
More Information
Set Membership: |
|

Aff3
Genomic Map