Assay Details
| Gene Symbol: Ednrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276296.1 | NP_001263225.1 | 7 - 8 | 1563 | 70 | |
| GenBank mRNA | AK076426.1 | - | 7 - 8 | 1557 | 70 | |
More Information
Set Membership: |
|

Ednrb
Genomic Map