Assay Details
| Gene Symbol: Esco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081222.1 | NP_001074691.1 | 6 - 7 | 2850 | 132 | |
| GenBank mRNA | AK040576.1 | - | 6 - 7 | 678 | 132 | |
| AK049589.1 | - | 5 - 6 | 566 | 132 | ||
| AK139549.1 | - | 6 - 7 | 2574 | 132 | ||
| AK145333.1 | - | 6 - 7 | 2851 | 132 | ||
| AK145870.1 | - | 5 - 6 | 498 | 132 | ||
| AK173297.1 | - | 4 - 5 | 2230 | 132 | ||
| BC008220.1 | - | 4 - 5 | 600 | 132 | ||
| BC076613.1 | - | 6 - 7 | 2580 | 132 | ||
| BC151069.1 | - | 6 - 7 | 2806 | 9087982 | 132 | |
| BC151077.1 | - | 6 - 7 | 2806 | 9087990 | 132 | |
| BC172153.1 | - | 6 - 7 | 2806 | 132 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Esco1
Genomic Map