Assay Details
| Gene Symbol: Esco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081222.1 | NP_001074691.1 | 8 - 9 | 3063 | 148 | |
| GenBank mRNA | AK040576.1 | - | 8 - 9 | 891 | 148 | |
| AK049589.1 | - | 7 - 8 | 779 | 148 | ||
| AK139549.1 | - | 8 - 9 | 2787 | 148 | ||
| AK145333.1 | - | 8 - 9 | 3064 | 148 | ||
| AK145870.1 | - | 7 - 8 | 711 | 148 | ||
| AK173297.1 | - | 6 - 7 | 2443 | 146 | ||
| BC076613.1 | - | 8 - 9 | 2793 | 148 | ||
| BC151069.1 | - | 8 - 9 | 3019 | 9087982 | 148 | |
| BC151077.1 | - | 8 - 9 | 3019 | 9087990 | 148 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Esco1
Genomic Map