Assay Details
| Gene Symbol: Rab18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278447.1 | NP_001265376.1 | 4 - 5 | 378 | 101 | |
| NM_181070.6 | NP_851415.1 | 4 - 5 | 360 | 101 | ||
| GenBank mRNA | AB232612.1 | - | 4 - 5 | 262 | 101 | |
| AK045514.1 | - | 4 - 5 | 323 | 101 | ||
| AK140901.1 | - | 4 - 5 | 328 | 101 | ||
| AK143673.1 | - | 4 - 5 | 336 | 101 | ||
| AK146177.1 | - | 4 - 5 | 285 | 101 | ||
| AK146397.1 | - | 4 - 5 | 285 | 101 | ||
| AK154920.1 | - | 4 - 5 | 337 | 101 | ||
| BC056351.1 | - | 4 - 5 | 314 | 6807284 | 101 | |
| L04966.1 | - | 4 - 5 | 266 | 101 | ||
| X80333.1 | - | 4 - 5 | 262 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab18
Genomic Map