Assay Details
| Gene Symbol: Thoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153552.3 | NP_705780.1 | 10 - 11 | 794 | 110 | |
| GenBank mRNA | AK031785.1 | - | 10 - 11 | 795 | 110 | |
| AK032200.1 | - | 10 - 11 | 795 | 110 | ||
| AK042867.1 | - | 10 - 11 | 800 | 110 | ||
| AK047991.1 | - | 6 - 7 | 2057 | 110 | ||
| AK052835.1 | - | 10 - 11 | 790 | 110 | ||
| AK139307.1 | - | 10 - 11 | 800 | 110 | ||
| AK167374.1 | - | 10 - 11 | 808 | 110 | ||
| BC027149.1 | - | 4 - 5 | 376 | 110 | ||
More Information
Set Membership: |
|

Thoc1
Genomic Map