Assay Details
| Gene Symbol: Thoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153552.3 | NP_705780.1 | 6 - 7 | 438 | 105 | |
| GenBank mRNA | AK031785.1 | - | 6 - 7 | 439 | 105 | |
| AK032200.1 | - | 6 - 7 | 439 | 105 | ||
| AK042867.1 | - | 6 - 7 | 444 | 105 | ||
| AK047991.1 | - | 2 - 3 | 1701 | 105 | ||
| AK052835.1 | - | 6 - 7 | 434 | 105 | ||
| AK139307.1 | - | 6 - 7 | 444 | 105 | ||
| AK167374.1 | - | 6 - 7 | 452 | 105 | ||
| AK168619.1 | - | 6 - 7 | 703 | 105 | ||
| AK215219.1 | - | 3 - 4 | 213 | 105 | ||
More Information
Set Membership: |
|

Thoc1
Genomic Map