Assay Details
| Gene Symbol: Sos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009231.2 | NP_033257.2 | 5 - 6 | 1252 | 153 | |
| GenBank mRNA | AK140310.1 | - | 5 - 6 | 1252 | 153 | |
More Information
Set Membership: |
|

Sos1
Genomic Map