Assay Details
| Gene Symbol: Sos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009231.2 | NP_033257.2 | 7 - 8 | 1499 | 95 | |
| GenBank mRNA | AK140310.1 | - | 7 - 8 | 1499 | 95 | |
More Information
Set Membership: |
|

Sos1
Genomic Map