Assay Details
| Gene Symbol: Arhgef33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145452.1 | NP_001138924.1 | 14 - 15 | 1753 | 97 | |
| GenBank mRNA | BE654086.1 | - | 1 - 2 | 139 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef33
Genomic Map