Assay Details
| Gene Symbol: Arhgef33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145452.1 | NP_001138924.1 | 17 - 18 | 2770 | 105 | |
| GenBank mRNA | AI848431.1 | - | 3 - 4 | 350 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P107482 | NM_001145452.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P107482 | AK053944.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673708 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 457169 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561100 | AK030112.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561100 | AK031245.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561100 | NM_178854.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 615834 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787374 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 591964 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678072 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 652227 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 116778 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737573 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 109749 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 189810 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749989 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 691570 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 610906 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 873184 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657055 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 536399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218377 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83358 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451341 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336580 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 103268 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 120101 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360115 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972263 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782428 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313186 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924362 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814978 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481970 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 332499 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 248637 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506007 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138697 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 388598 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 152352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884848 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 268692 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 21344 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 866971 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 602186 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471046 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55252 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412781 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 408773 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350024 | AK132237.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350024 | AK141053.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350024 | AY288425.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350024 | BC062104.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350024 | BC114990.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350024 | BC115824.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350024 | BC148305.2 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 350024 | NM_181752.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467207 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649023 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 897801 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 488522 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 632127 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 100969 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 133509 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 150800 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 90379 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 112389 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 62103 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 115028 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 1772 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 89229 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 152646 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P124684 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153065 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P107482 | NM_001145452.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P107482 | AK053944.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P124684 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08864781 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08864693 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153065 | - | - |
| Whole Mouse Genome G4122A | A_52_P228794 | AK035848.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P228794 | AK141710.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P228794 | AK163885.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P228794 | AK047849.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P228794 | NM_009231.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153065 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P124684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382541 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382553 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382539 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382738 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13301030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13381390 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382535 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382556 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13301321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382490 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13474486 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382528 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382521 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13211085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13381396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13303383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13098100 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13342617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13249244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13220051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13553921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382492 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13381395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382726 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382656 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382719 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13301326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13246414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13185741 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382561 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382491 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13258634 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13412989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382529 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382504 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13189533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382658 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13363296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13569712 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13211086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31382557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13186633 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10447110 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447112 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447109 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447111 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447114 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447113 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447117 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447405_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436410_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1458753_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10447108 | - | - |
| Mouse Genome 430 2.0 Array | 1458753_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436410_at | - | - |
| Mouse Genome 430 2.0 Array | 1458753_at | - | - |
| Mouse Genome 430 2.0 Array | 1436410_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700011396.mm.1 | NM_001145452.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700011396.mm.1 | NM_001145452.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700011396.mm.1 | TC1700001215.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1458753_at | - | - |
| Mouse Expression Set 430 | 1447405_PM_at | - | - |
| Mouse Expression Set 430 | 1447405_at | - | - |
| Mouse Expression Set 430 | 1458753_PM_at | - | - |
| Mouse Expression Set 430 | 1436410_at | - | - |
| Mouse Expression Set 430 | 1436410_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458753_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1458753_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436410_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436410_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 436825 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1241803 | - | - |

Arhgef33
Genomic Map