Assay Details
| Gene Symbol: Cdkl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033443.4 | NP_001028615.1 | 2 - 3 | 335 | 141 | |
| GenBank mRNA | AK157995.1 | - | 2 - 3 | 336 | 141 | |
| BC138988.1 | - | 1 - 2 | 233 | 8862010 | 141 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdkl4
Genomic Map