Assay Details
| Gene Symbol: Eif1ad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027236.2 | NP_081512.1 | 3 - 4 | 718 | 101 | |
| GenBank mRNA | AK008070.1 | - | 3 - 4 | 528 | 101 | |
| AK153194.1 | - | 3 - 4 | 549 | 101 | ||
| AK158977.1 | - | 3 - 4 | 536 | 101 | ||
| AK168102.1 | - | 3 - 4 | 522 | 101 | ||
| AK168253.1 | - | 3 - 4 | 522 | 101 | ||
| AK168598.1 | - | 3 - 4 | 545 | 101 | ||
| BC024640.1 | - | 3 - 4 | 718 | 4038046 | 101 | |
More Information
Set Membership: |
|

Eif1ad
Genomic Map