Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00017946.mm.1 |
NM_001190409.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017946.mm.1 |
NM_001190409.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017946.mm.1 |
NM_025952.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017946.mm.1 |
NM_025952.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017946.mm.1 |
TC0X00002825.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00017944.mm.1 |
NM_001190409.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017944.mm.1 |
NM_001190409.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017944.mm.1 |
NM_025952.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017944.mm.1 |
NM_025952.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017944.mm.1 |
TC0X00002825.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00017943.mm.1 |
NM_001190409.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017943.mm.1 |
NM_001190409.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017943.mm.1 |
NM_025952.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017943.mm.1 |
NM_025952.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00017943.mm.1 |
TC0X00002825.mm.1 |
MTA TC |