Assay Details
| Gene Symbol: Scyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023912.2 | NP_076401.1 | 6 - 7 | 1086 | 72 | |
| GenBank mRNA | AF276514.1 | - | 6 - 7 | 1086 | 72 | |
| AK019240.1 | - | 6 - 7 | 1079 | 72 | ||
| AK134356.1 | - | 6 - 7 | 1085 | 72 | ||
| AK144134.1 | - | 6 - 7 | 1072 | 72 | ||
| AK145869.1 | - | 6 - 7 | 1106 | 72 | ||
| AK161813.1 | - | 6 - 7 | 1085 | 72 | ||
| AK200223.1 | - | 1 - 2 | 77 | 72 | ||
| AK219353.1 | - | 1 - 2 | 77 | 72 | ||
| BC034519.1 | - | 6 - 7 | 1072 | 3672524 | 72 | |
| HM856325.1 | - | 6 - 7 | 1086 | 72 | ||
| HM856326.1 | - | 6 - 7 | 1086 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scyl1
Genomic Map